Learn More
Hirschmann™ rea-stabil™ Liquid Bottle
Product Code. 10379561
Change view
Click to view available options
Volume (Metric):
200 mL
Unit Size:
200mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10379561 200 mL 200mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10379561 Supplier Hirschmann™ Supplier No. 9780100
  • ONLINE EXCLUSIVE PRICE
    98.89€
    79.00€ / 200mL
    Save 19.89€
    20% Off
 Restricted Item
In Stock
Helps avoid algae, bacteria and mould formation in the laboratory. Hirschmann Instruments™ rea-stabil™ Liquid Bottle is for use with hot water baths, thermostats and similar areas of use.
  • Bactericide and fungicide
  • Non-toxic
  • Very economical - 0.1% working concentration
Specifications
Product Type Stabilization Concentrate
Form Liquid
Concentration 0.1%
Container Type Bottle
Volume (Metric) 200 mL
compliance-icons
Product Identifier
  • rea-stabil
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.