missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBP4/Retinol-Binding Protein 4 Rat anti-Human, Mouse, Alexa Fluor 350, Clone: 423619, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB34761U-100UG
This item is not returnable.
View return policy
Description
RBP4/Retinol-Binding Protein 4 Monoclonal specifically detects RBP4/Retinol-Binding Protein 4 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
| RBP4/Retinol-Binding Protein 4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_035385 | |
| RBP4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse RBP4/Retinol-Binding Protein 4 Met1-Leu201 Accession # NP_035385 | |
| RUO | |
| 5950 | |
| Human, Mouse | |
| IgG2a |
| Flow Cytometry | |
| 423619 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, RetinolBinding Protein 4, retinol-binding protein 4, plasma | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse RBP4/Retinol-Binding Protein 4 in direct ELISA and Western Blot. In Western blots, detects human RBP4/Retinol-Binding Protein 4. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction