Learn More
Novus Biologicals™ RBP4/Retinol-Binding Protein 4 Antibody Pair [HRP]
Product Code. 18266567 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
Product Code. 18266567
Brand: Novus Biologicals™ NBP27958315PLATES
Mouse Monoclonal Antibody Pair
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human RBP4/Retinol-Binding Protein 4.
TRUSTED_SUSTAINABILITY
Specifications
Assay Range 23.44 pg/mL
Assay Sensitivity 23.44-1500 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human RBP4/Retinol-Binding Protein 4.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, retinol-binding protein 4, plasma
Gene ID (Entrez) 5950
Gene Symbol RBP4
Host Species Mouse
Kit Contents
  • Detection Antibody: 0.2 mg/mL of mouse anti-Human RBP4/Retinol-Binding Protein 4 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 1 mg/mL of mouse anti-Human RBP4/Retinol-Binding Protein 4 monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 25 ng of recombinant Human RBP4/Retinol-Binding Protein 4.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cancer, Diabetes Research, Lipid and Metabolism
    Storage Requirements Storage is content dependent.
    Target RBP4/Retinol-Binding Protein 4
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1500 pg/mL is recommended.