Learn More
Novus Biologicals™ Raw 264.7 Nuclear Cell Lysate
Product Code. 18518680 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18518680 0.05 mg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18518680 Supplier Novus Biologicals™ Supplier No. L024V3
 Restricted Item
Estimated Shipment: 14-10-2026
Nuclear Extract Lysate
TRUSTED_SUSTAINABILITY
Specifications
Preparation Method The lysate is ready to load on SDS-PAGE for Western blotting.
Quantity 0.05 mg
Content And Storage Store at -80C. Avoid freeze-thaw cycles.
Target Raw 264.7