Learn More
Novus Biologicals™ Raw 264.7 Hypoxic/ Normoxic Cell Lysate
Product Code. 18691699 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18691699 2 Vials Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18691699 Supplier Novus Biologicals™ Supplier No. NBP311860
  • ONLINE EXCLUSIVE PRICE
    382.00€
    361.20€ / Pack of 2
    Save 20.80€
    5% Off
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
Add to basket
Cell Lysate
TRUSTED_SUSTAINABILITY
Specifications
Preparation Method RAW264.7 whole cell lysate is provided as a Western blot positive control. The lysates are provided as a set of treated and untreated samples. Use 10 μL / lane for a standard mini-gel blot (1.0 mg/mL).
Quantity 2 Vials
Host Species Mouse
Content And Storage Store at -20°C to -80°C. Avoid freeze/thaw cycles.
Target Raw 264.7