Learn More
R&D Systems™ Rat Neuroligin 1/NLGN1 (NP_446320.1) VersaClone cDNA
Specifications
Specifications
| Gene | Neuregulin-1/NRG1 |
| Accession Number | NP_446320.1 |
| Gene ID (Entrez) | 3084 |
| Gene Alias | ARIA, GGF2, GGFglial growth factor, Heregulin-1, HGL, HGLneu differentiation factor, HRG, HRG1, HRG1-alpha, HRG1-beta 1, HRGAneuregulin 1 type IV beta 1 A, MST131, MSTP131, NDFheregulin, alpha (45 kD, ERBB2 p185-activator), neuregulin 1, neuregulin 1 type IV beta 3, Neuregulin1, NRG1, pro-neuregulin-1, membrane-bound isoform, pro-NRG1, sensory and motor neuron derived factor, SMDF |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Rat |
| Quantity | 10 μg |