Learn More
R&D Systems™ Rat Luminex™ Discovery Assay
Specifications
Specifications
| Quantity | 11 Plex |
| Species | Rat |
| Binding Target | Multiplex Cytokine Assay Kits |
| Content And Storage | Store the unopened kit at 2°C to 8°C. Do not use past the expiration date on the label. Avoid freezing microparticles. Protect microparticles from light. |