Learn More
R&D Systems™ Rat Luminex™ Discovery Assay
Click to view available options
Quantity:
1 Plex
11 Plex
2 Plex
4 Plex
8 Plex
Unit Size:
1 set
Each
Specifications
Specifications
| Quantity | 2 Plex |
| Species | Rat |
| Binding Target | Multiplex Cytokine Assay Kits |
| Content And Storage | Store the unopened kit at 2°C to 8°C. Do not use past the expiration date on the label. Avoid freezing microparticles. Protect microparticles from light. |