missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat Lipocalin-2/NGAL DuoSet ELISA

Product Code. 30021046
Change view
Click to view available options
Quantity:
15 x 96-well Plates
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30021046 15 x 96-well Plates 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30021046 Supplier R&D Systems™ Supplier No. DY3508

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Lipocalin-2/NGAL DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Rat Lipocalin-2/NGAL DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies rat Lipocalin-2/NGAL.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 15 x 96-well Plates
Conjugate Biotin
Assay Range 78.10 to 5000 pg/mL
For Use With (Application) ELISA
Gene ID (Entrez) 170496
Synonym 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin2, Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGAL, NGALlipocalin-2, Oncogene 24p3, p25, Siderocalin, Uterocalin
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.