Learn More
Novus Biologicals™ Rat IgG ELISA Kit (Chemiluminescence)
Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Specifications
Specifications
| Assay Range | 7.81 to 500 pg/mL |
| Assay Sensitivity | 4.69 pg/mL |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA Kit (Chemiluminescence) |
| Sample Type | Serum, Plasma, Biological Fluids |
| Specificity | This kit recognizes Rat IgG in samples. No significant cross-reactivity or interference between Rat IgG and analogues was observed. |
| Target Species | Rat |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3500 |
| Gene Symbol | IGHG1 |
| Show More |