Learn More
Abcam Rat heart tissue lysate - mitochondrial extract, 2 mg
Product Code. 32143809 Shop All Abcam Products
Change view
Click to view available options
Unit Size:
2000µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
32143809 2000µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32143809 Supplier Abcam Supplier No. ab1103472mg
 Restricted Item
Estimated Shipment: 09-12-2026
Rat heart cell/tissue lysate.
  • Species: Rat
  • Tissue: heart
  • Form: Liquid
Specifications
For Use With (Application) Western Blot, SDS-PAGE, Immunoprecipitation