Learn More
R&D Systems™ Rat CD8 beta (CAA27850.1) VersaClone cDNA
Specifications
Specifications
| Gene | CD8 beta |
| Accession Number | CAA27850.1 |
| Concentration | 10 μg (at 0.2 μg/μL) |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 640 bp |
| Gene ID (Entrez) | 926 |
| Gene Alias | CD8 antigen, beta polypeptide 1 (p37), CD8 beta, CD8b, CD8b antigen, CD8b molecule, CD8B1, CD8B1P37, LEU2, LY3, LYT3, MGC119115, P37, T lymphocyte surface glycoprotein beta chain, T-cell surface glycoprotein CD8 beta chain |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Rat |
| Product Line | R&D Systems |
| Show More |