missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAGE Rat anti-Mouse, Rat, Clone: 175410, R&D Systems™

Rat Monoclonal Antibody has been used in 15 publications
Brand: R&D Systems MAB1179-500
This item is not returnable.
View return policy
Description
AGER Monoclonal specifically detects AGER in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| RAGE | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| NP_031451 | |
| Ager | |
| Mouse myeloma cell line NS0-derived recombinant mouse RAGE Gly23-Ala342 Accession # NP_031451 | |
| 500 μg | |
| Primary | |
| Detects mouse and rat RAGE in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human RAGE is observed. | |
| Mouse, Rat | |
| Supernatant |
| Western Blot, Immunohistochemistry | |
| 175410 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| advanced glycosylation end product-specific receptor, AGER, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products, SCARJ1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5891 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction