Learn More
Rat anti-Mouse IgG2b, Clone: R9-91, Secondary Antibody, BD
Brand: BD Biosciences 553396
Additional Details : Weight : 0.01000kg
Description
The R9-91 antibody recognizes an epitope in the CH3 domain of mouse IgG2b of Igh-C[a] and Igh-C[b] haplotypes. It does not react with other Ig isotypes. Detection of surface immunoglobulin on B lymphoma cells has been demonstrated with R9-91 mAb
Host Species: Rat
Clone: R9-91
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Pooled Mouse Ig
ELISA, Flow Cytometry
Specifications
Rat anti-Mouse IgG2b | |
R9-91 | |
Rat | |
0.5 mg | |
Immunology | |
Mouse |
Monoclonal | |
0.5mg/mL | |
Affinity Purified | |
RUO | |
Secondary | |
IgG1 |