Learn More
Novus Biologicals™ RASGEF1B 293T Cell Transient Overexpression Lysate
Product Code. 18555602 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18555602 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18555602 Supplier Novus Biologicals™ Supplier No. H00153020T01
 Restricted Item
Estimated Shipment: 17-08-2026
Specifications
Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Research Category Signal Transduction
Target RASGEF1B
Synonym RasGEF domain family, member 1 B, ras-GEF domain-containing family member 1 B
Gene Symbol RASGEF1B