Learn More
Ras Rat anti-Human, Clone: 342404, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems MAB3429-SP
Encompass
Product Code. 15716224
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
Ras Monoclonal specifically detects Ras in Human samples. It is validated for Western Blot.