Learn More
Novus Biologicals™ Rapid Tissue Preservation Solution
Product Code. 18649580 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18649580 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18649580 Supplier Novus Biologicals™ Supplier No. NBP312177
 Restricted Item
Estimated Shipment: 23-10-2026
Specifications
Content And Storage Store at room temperature.
Quantity 10 mL
For Use With (Application) Immunohistochemistry, Immunohistochemistry-Paraffin