missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Raphin 1

Description
Raphin 1 is an inhibitor of the regulatory subunit PPP1R15B of protein phosphatase (Kd = 33 nM). Displays >30-fold selectivity for PPP1R15B over PPP1R15A. Inhibits PPP1R15B-mediated dephosphorylation and protein synthesis in vitro. Induces transient reduction in protein synthesis in brains of wild type mice after single dose. Decreases SDS-insoluble huntingtin assemblies and nuclear inclusions in a mouse model of Huntington's disease. Orally bioavailable and brain penetrant.
Specifications
Specifications
| Chemical Name or Material | (2E)-2-[(2,3-Dichlorophenyl)methylene]hydrazinecarboximidamide |
| CAS | 2022961-17-5 |
| Quantity | 10 mg |
| Molecular Formula | C8H8Cl2N4 |
| Formula Weight | 231.08 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?