Learn More
Ranitidine, TRC
Specifications
Specifications
| Chemical Name or Material | Ranitidine |
| CAS | 66357-35-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C13 H22 N4 O3 S |
| Quantity | 50 mg |
| Synonym | 1,1-Ethenediamine','N-[2-[[[5-[(dimethylamino)methyl]-2-furanyl]methyl]thio]ethyl]-N'-methyl-2-nitro-, N-[2-[[[5-[(Dimethylamino)methyl]-2-furanyl]methyl]thio]ethyl]-N'-methyl-2-nitro-1,1-ethenediamine, Label, Midaven, Ranitidine, Ulcerit, Zenetac |
| InChI Formula | InChI=1S/C13H22N4O3S/c1-14-13(9-17(18)19)15-6-7-21-10-12-5-4-11(20-12)8-16(2)3/h4-5,9,14-15H,6-8,10H2,1-3H3 |
| SMILES | CNC(=C[N+](=O)[O-])NCCSCc1oc(CN(C)C)cc1 |
| IUPAC Name | 1-N'-[2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine |
| Molecular Weight (g/mol) | 314.4 |
| Show More |