Learn More
Emma Safety Footwear Ranger Nubuck Leather Outdoor D-Fit Shoes
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Each
Description
- Nubuck/Cordura™ uppers
- Hydro-Tec™ sanitized silver lining
- Non-woven antistatic insole
- GripForce™ Easy Twist outsole
Specifications
Specifications
| Color | Gray |
| Liner Material | Hydro-Tec sanitized silver lining |
| Material | Leather |
| Size | 48 |
| Sole Features | Non-woven antistatic insole |
| Sole Material | GripForce™ Easy Twist outsole |
| Toe Style | Steel toe |
| Product Type | Nubuck Leather Outdoor Shoes |