Learn More
Ohaus™ Ranger™ Count 3000 Multipurpose Compact Counting Scale
Product Code. p-7158834 Shop All Ohaus Products
Change view
Click to view available options
Capacity:
1.5 kg
3.0 kg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
15853595 1.5 kg Each
13445119 3.0 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15853595 Supplier Ohaus™ Supplier No. 30060907
Compact, multipurpose counting scale with weighing, parts counting, check counting, and accumulation functions.
  • Precise counting and weighing performance with 30 item library
  • The most user-friendly counting scale on the market
  • Rapid stabilization provides unmatched throughput
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 210 Hours with Rechargeable Battery
Connection Type Ethernet (Available as an Accessory)USB (Available as an Accessory)RS232 (Included)2nd RS232 (Available as an Accessory)
Construction ABS housing, stainless steel platform, numeric keypad, menu lock switch, up front level indicator, transportation handle
Capacity 1.5 kg
Display Type 3-window Backlight LCD + 3 LED
Readability 0.500000 g
Includes RS232, AC power, rechargeable battery
Linearity 0.100000 g
Material Stainless Steel Platform
Model RC31P1502-M
Repeatability 0.100000 g
Dimensions (L x W) 311.00 x 327.00 mm
Dimensions (L x W x H) 311.00 x 327.00 x 117.000 mm
Length (English) 12.20 in.
Length (Metric) 311.00 mm
Length (Metric) Pan 225 mm
Width (English) 12.90 in.
Width (Metric) 327.00 mm
Width (Metric) Pan 300 mm
Height (English) 4.60 in.
Height (Metric) 117.000 mm
Depth (English) 12.20 in.
Depth (Metric) 311.00 mm
Electrical Requirements 100/240 VAC 50/60 Hz internal universal power supply, internal rechargeable lead acid battery