Learn More
Ohaus™ Explorer™ Precision High Capacity Balance
Product Code. 14844721 Shop All Ohaus Products
Click to view available options
:
Capacity: 30kg
Product Code. 14844721
Brand: Ohaus™ 30078074
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Combine modern features and design elements to offer unmatched functionality in high-performance balances. Ohaus™ Explorer™ Precision High Capacity Balance simplify even the most complex laboratory measurements.
Ohaus™ Explorer™ has advanced applications to simplify even the most complex laboratory measurements. Whether it's determining the difference between initial and residual weights or calculating the density of solids and liquids, the Explorer™ eliminates the need for time consuming manual calculations and data logging. The high resolution display and innovative user interface make balance setup and application use effortless. All 3 decimal place balances come with glass draft shield.
  • High resolution multi-lingual color VGA touch display with icons for simple operation
  • Programmable multi-functional touch free sensors for cleanliness, speed and safety
  • Multiple application modes plus library for storage of personal application settings
TRUSTED_SUSTAINABILITY
Specifications
Capacity 35 kg
Display Type Full-color 5.7 in. VGA, Touchscreen
Readability 0.1 g
Linearity ±0.2 g
Min. Sample Weight 210 g
Model 3000
Repeatability 0.1 g
Stabilization Time 1 sec.
Type Analytical Balance
Dimensions (L x W x H) 443 x 377 x 120 mm
Diameter (Metric) Outer Pan 377 x 311 mm
Electrical Requirements AC Power (Included); Rechargeable Battery (Not Included)