Learn More
R&D Systems™ Polystyrene Microplates
Product Code. p-4758030
Click to view available options
Color:
Clear
Unit Size:
Pack of 25
Product Code. 30117142
Brand: R&D Systems™ DY990
high-binding, flat-bottom polystyrene microplates
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 8 x 12
Color Clear
Product Type ELISA Microplate
Material Polystyrene
Volume (Metric) Well 360 μL
No. of Wells 96
Well Shape Flat Bottom (F-well)
For Use With (Application) ELISA