Learn More
Raloxifene-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Raloxifene-d4 |
| CAS | 1185076-44-1 |
| Molecular Formula | C28 2H4 H23 N O4 S |
| Quantity | 10 mg |
| Synonym | [6-Hydroxy-2-(4-hydroxyphenyl)-1-benzothiophen-3-yl]-[4-(1,1,2,2-tetradeuterio-2-piperidin-1-ylethoxy)phenyl]methanone, 2-(4-Hydroxyphenyl)-3-[4-[2-(piperidin-1-yl)ethoxy-d4]benzoyl]-1-benzothiophen-6-ol, [6-Hydroxy-2-(4-hydroxyphenyl)-1-benzothiophen-3-yl]-[4-(2-piperidin-1-ylethoxy-1,1,2,2-d4)phenyl]methanone, Raloxifene-d4, Raloxifene-D4 (ethoxy-1,1,2,2-D4) |
| InChI Formula | InChI=1S/C28H27NO4S/c30-21-8-4-20(5-9-21)28-26(24-13-10-22(31)18-25(24)34-28)27(32)19-6-11-23(12-7-19)33-17-16-29-14-2-1-3-15-29/h4-13,18,30-31H,1-3,14-17H2/i6D,7D,11D,12D |
| SMILES | [2H]c1c([2H])c(C(=O)c2c(sc3cc(O)ccc23)c4ccc(O)cc4)c([2H])c([2H])c1OCCN5CCCCC5 |
| IUPAC Name | [6-hydroxy-2-(4-hydroxyphenyl)-1-benzothiophen-3-yl]-[2,3,5,6-tetradeuterio-4-(2-piperidin-1-ylethoxy)phenyl]methanone |
| Molecular Weight (g/mol) | 477.61 |
| Formula Weight | 477.1912 |