missing translation for 'onlineSavingsMsg'
Learn More

Raloxifene-d4 4’-Glucuronide, TRC

Product Code. 30421769
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30421769 5 mg 5mg
30422631 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30421769 Supplier Toronto Research Chemicals Supplier No. TRCR1000225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C34 D4 H31 N O10 S
Molecular Weight (g/mol) 653.732
Synonym 4-[6-Hydroxy-3-[4-[2-(1-piperidinyl)ethoxy-d4]benzoyl]benzo[b]thien-2-yl]phenyl-beta-D-glucopyranosiduronic Acid
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[4-[6-hydroxy-3-[4-(1,1,2,2-tetradeuterio-2-piperidin-1-ylethoxy)benzoyl]-1-benzothiophen-2-yl]phenoxy]oxane-2-carboxylic acid
SMILES [2H]C([2H])(Oc1ccc(cc1)C(=O)c2c(sc3cc(O)ccc23)c4ccc(O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)O)cc4)C([2H])([2H])N6CCCCC6

Tekniske data

Quantity 5 mg
InChI Formula InChI=1S/C34H35NO10S/c36-21-8-13-24-25(18-21)46-32(20-6-11-23(12-7-20)44-34-30(40)28(38)29(39)31(45-34)33(41)42)26(24)27(37)19-4-9-22(10-5-19)43-17-16-35-14-2-1-3-15-35/h4-13,18,28-31,34,36,38-40H,1-3,14-17H2,(H,41,42)/t28-,29-,30+,31-,34+/m0/s1/i16D2,17D2
Formula Weight 653.223
Chemical Name or Material Raloxifene-d4 4’-Glucuronide
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.