Learn More
Uvex™ Rain Jacket 7491-Yellow
Product Code. p-7231970 Shop All Uvex Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Each
Product Code. 17240352
Brand: Uvex™ 8831009
Only null left
Add to basket
Add to basket
PU rainwear offers moisture protection with optimum wearer comfort. Incredibly malleable and tear resistant the ultra-lightweight material for use in the wind and rain.
  • Rain jacket
  • Material: 100% polyester with PU coating 170 gsm
  • Color: yellow
  • Long jacket with adjustable hood, adjustable hem, elastic sleeve hem
  • Front flap with zipper and press button, two side pockets and flap, comfortable to wear
  • Sizes: S-3XL
  • Oversize surcharge from Gr. XXL + 10% do not wash, do not bleach, do not dry, do not iron, do not clean
TRUSTED_SUSTAINABILITY
Specifications
Size Small
Gender Men's
Material Polyester
Color Yellow
Closure Type Zipper
Disposable Reusable
Product Type Rain Jacket