missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAGE Rat anti-Mouse, Alexa Fluor 488, Clone-697023, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems FAB11795G-100UG
This item is not returnable.
View return policy
Description
AGER Monoclonal specifically detects AGER in Mouse samples. It is validated for Flow Cytometry.Specifications
| RAGE | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.5 ug/10^6 cells | |
| NP_031451 | |
| Ager | |
| Mouse myeloma cell line NS0-derived recombinant mouse RAGE Gly23-Leu342 Accession # NP_031451 | |
| 100 μg | |
| Primary | |
| Detects mouse RAGE in direct ELISAs and Western blots. In Western blots, approximately 15% cross-reactivity with recombinant canine RAGE and no cross-reactivity with recombinant human RAGE or recombinant rat RAGE is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry | |
| 697023 | |
| Alexa Fluor 488 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| advanced glycosylation end product-specific receptor, AGER, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products, SCARJ1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5891 | |
| Mouse | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction