Learn More
TFA Dostmann™ Radio-Thermo-Hygrometer
Product Code. 10810131
Click to view available options
Includes:
Thermometer, hygrometer
Unit Size:
Each
Product Code. 10810131
Brand: TFA Dostmann™ 30.3013.IT
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Humidity Range 1 to 99%
Temperature Range (Metric) -40°C to +60°C (Outdoor), -10°C to +60°C (Indoor)
Battery Type 3 x 1.5 V AA for display unit; 2 x 1.5 V AA for transmitter
Mounting Hanging or standing
Includes Thermometer, hygrometer