Learn More
Fisherbrand™ Radiation Safety Gamma Boxes
Product Code. 12357117 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Height (Metric):
130mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
12357117 130mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12357117
Estimated Shipment: 06-04-2027
This gamma radiation bin is ideal for holding microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes.
  • Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts to hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
  • Also available is our range of floor-standing and benchtop bins with anti-slip feet and hinged lids, which serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Both the Beta and Gamma storage bins are available in five sizes, and the two largest bin models have wheels for easy transportation
  • Optional heavy duty drawstring bags may also be purchased
Specifications
For Use With (Equipment) CSR-BAG1
Height (Metric) 130mm
Tube Size 13mm x 13mm x 13mm
Width (Metric) 130mm
Capacity 2 L