Learn More
Thermo Scientific™ RadEye™ G20-10 and G20-ER10 X-Ray and Gamma Survey Meters
Product Code. 11960737 Shop All Thermo Scientific Products
Change view
Click to view available options
Dose Rate Limit:
2 mSv/h (200 mrem/h)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dose Rate Limit Unit Size
11960737 2 mSv/h (200 mrem/h) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11960737 Supplier Thermo Scientific™ Supplier No. 4250687
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Protect nuclear medicine personnel and nuclear facility employees with gamma survey meters for continuous monitoring of external or ambient ionizing radiation fields.
Nuclear medicine personnel who work with medical isotopes or in facilities that use radiation need to monitor their risk of exposure at all times. Protect employees and meet occupational safety regulations with the Thermo Scientific™ RadEye G20-10 and G20-ER10 X-ray and Gamma Survey Meters. These lightweight, compact Geiger counters are ideal for ambient equivalent dose and dose rate measurement, with a flat energy response curve from 17keV to 1.3MeV according to ambient equivalent dose H*(10). The RadEye G20-ER10 model covers the whole dose rate range from background to 100 mSv/h.
RadEye G20-10 / G20-ER10 X-Ray and Gamma Survey Meters for ambient equivalent dose and dose rate measurement are the perfect Geiger counters for nuclear industry personnel as well as civil defence, fire brigades, and hospital and pharmaceutical industry employes who work with radiation-emitting equipment.
  • Fast response, even below 1 μSv/h.
  • Rugged, compact design with thick rubber protective cover.
  • Menu-driven user interface for low training cost and immediate familiarity.
  • Huge internal data memory for both scaler results and continuous data recording.
  • Bright backlit LCD display, offering plain text messages in multiple language choices.
  • Audible indication: single pulse or chirper mode proportional to count rate.
  • Earphone output for operation in loud environments.
TRUSTED_SUSTAINABILITY
Specifications
Sensitivity 0.01μSv/h to2 mSv/h
Certifications/Compliance Designed to meet and exceed IEC 60846-1.
Description RadEye G20-10 X-Ray and Gamma Survey Meters, Upper dose rate limit: 2 mSv/h
Energy Range 17 keV to 3 MeV( ± 30%); 17 keV to 3.0 MeV according to IEC 60846-1 (2009)
Display Type Backlit LCD
Battery Type 2 AAA
Product Type Survey Meter
Measurement Range 0.01 μSv/h to 2 mSv/h
Operating Temperature (Metric) −20°C to +50°C
Height (English) 5.12 in.
Operating Temperature (English) -4°F to 122°F
Response Time Approx. 4 cps per µSv/h
Detector Type Pancake GM with multi-element filter
Linearity +/- 10% within the measurement range
Width (Metric) 6.7 cm
Weight (Metric) 0.3 kg
For Use With (Application) Civil defense, fire brigades, hospitals, nuclear industry, and pharmaceutical industry
Radiation Type Gamma, X-Ray
Units Sv/h; rem/h
Depth (Metric) 62 mm
Width (English) 2.64 in.
Battery Life >500 hr.s on two AAA batteries
Dose Rate Limit 2 mSv/h (200 mrem/h)
Weight (English) 0.66 lb.
Depth (English) 2.44 in.
Height (Metric) 13 cm