missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ RAD51AP1 293T Cell Transient Overexpression Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.01mL
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Research Category | DNA Repair, Homologous Recombination |
| Target | RAD51AP1 |
| Synonym | PIR51RAD51-interacting protein, RAD51 associated protein 1, RAD51-associated protein 1 |
| Gene Symbol | RAD51AP1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction