Learn More
Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer
Product Code. 11704908 Shop All Thermo Scientific Products
Click to view available options
Holds:
10 Canisters (for cord blood, Model 4000610)
161 1.2 mL/2 mL Ampules
161 4 mL/5 mL Ampules
162 Canes, 5 to 6 Ampules/Cane
3 x 4/5 mL Cryo Racks (Large)
3 x 4/5 mL Cryo Racks (Small)
40 0.25/0.5 mL Straws
48 Skin Packets (2 level)
5 x 1.2/2 mL Cryo Racks (Large)
5 x 1.2/2 mL Cryo Racks (Small)
76 1.2 mL/2 mL Ampules
76 4 mL/5 mL Ampules
77 Canes
Adjustable, 10-position
Type:
Cane Freezing Rack
Canister Rack
Skin Rack
Straw Rack
Vial Rack (Large)
Vial Rack (Small)
Vial Rack Holder
Unit Size:
Each
Product Code. 11704908
Brand: Thermo Scientific™ 4000706
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer.
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer. Includes an assortment of cane, canister, straw and vial racks.
  • Warranty varies by country, please contact us for details
TRUSTED_SUSTAINABILITY
Specifications
Color Silver
Description Three 4 mL/5 mL Cryo Racks (Small)
For Use With (Equipment) All CryoMed controlled-rate freezers
Holds 3 x 4/5 mL Cryo Racks (Small)
Type Vial Rack Holder