Learn More
Thermo Scientific™ Racks and Inserts for Refrigerated and Heated Bath Circulators
Product Code. 10048294 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Rack
Rack Insert
Holds:
100-10 mm Tubes
13-25 mm Tubes
20-16 mm Tubes
39-10 mm Tubes
60-16 mm Tubes
8-25 mm Tubes
Depends on insert purchased
Dishes, plates, bags, pouches
For Use With (Equipment):
A25B, A40, S21, S30
A5B,A10B A24B,S49,S19T,S14P,S21P
S13, S12T, S15
Unit Size:
Each
Pack of 2
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Holds For Use With (Equipment) unitSize
10048294 Rack Depends on insert purchased A5B,A10B A24B,S49,S19T,S14P,S21P Each
11620961 Rack Insert 100-10 mm Tubes A5B,A10B A24B,S49,S19T,S14P,S21P Each
11960715 Rack Insert 60-16 mm Tubes A5B,A10B A24B,S49,S19T,S14P,S21P Each
10458434 Rack Insert Dishes, plates, bags, pouches A5B,A10B A24B,S49,S19T,S14P,S21P Each
11977274 Rack Depends on insert purchased A25B, A40, S21, S30 Each
11907284 Rack Insert 13-25 mm Tubes A25B, A40, S21, S30 Pack of 2
11917284 Rack Insert Dishes, plates, bags, pouches A25B, A40, S21, S30 Pack of 2
11680981 Rack Depends on insert purchased S13, S12T, S15 Each
11927284 Rack Insert 39-10 mm Tubes S13, S12T, S15 Pack of 2
11668008 Rack Insert 20-16 mm Tubes S13, S12T, S15 Each
11947284 Rack Insert 8-25 mm Tubes S13, S12T, S15 Pack of 2
11957284 Rack Insert Dishes, plates, bags, pouches S13, S12T, S15 Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 10048294 Supplier Thermo Scientific™ Supplier No. 1600002
Only null left
Add to basket
Add to basket
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) A5B,A10B A24B,S49,S19T,S14P,S21P
Type Rack
Holds Depends on insert purchased
Material Stainless Steel