Learn More
rac-Hesperetin-d3, TRC
Specifications
Specifications
| Chemical Name or Material | rac-Hesperetin-d3 |
| CAS | 1346605-26-2 |
| Molecular Formula | C16H11D3O6 |
| Quantity | 5 mg |
| Synonym | 2,3-Dihydro-5,7-dihydroxy-2-[3-hydroxy-4-(methoxy-d3)phenyl)-4H-1-benzopyran-4-one, 3',5,7-Trihydroxy-4'-(methoxy-d3)flavanone, Eriodictyol-d3 4'-Monomethyl Ether |
| InChI Formula | InChI=1S/C16H14O6/c1-21-13-3-2-8(4-10(13)18)14-7-12(20)16-11(19)5-9(17)6-15(16)22-14/h2-6,14,17-19H,7H2,1H3/i1D3 |
| SMILES | OC1=CC(O)=C(C(CC(C2=CC(O)=C(OC([2H])([2H])[2H])C=C2)O3)=O)C3=C1 |
| IUPAC Name | 5,7-dihydroxy-2-(3-hydroxy-4-(methoxy-d3)phenyl)chroman-4-one |
| Molecular Weight (g/mol) | 305.3 |
| Formula Weight | 305.1 |