Learn More
rac Epinephrine-d3, TRC
Product Code. 30368849
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368849 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30368849 Supplier Toronto Research Chemicals Supplier No. TRCE588582100MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (±)-Epinephrine-D3 (Adrenaline)
CAS 1189977-29-4
Purity Grade Notes HPLC
Molecular Formula C9 2H3 H10 N O3
Quantity 100 mg
Synonym rac-Epinephrine-d3, 4-[1-Hydroxy-2-(trideuteriomethylamino)ethyl]benzene-1,2-diol, 2-[(Methyl-d3)amino]-1-(3,4-dihydroxyphenyl)ethanol, DL-Adrenaline-d3 (N-methyl-d3), dl-Epinephrine-d3
InChI Formula InChI=1S/C9H13NO3/c1-10-5-9(13)6-2-3-7(11)8(12)4-6/h2-4,9-13H,5H2,1H3/i1D3
SMILES [2H]C([2H])([2H])NCC(O)c1ccc(O)c(O)c1
IUPAC Name 4-[1-hydroxy-2-(trideuteriomethylamino)ethyl]benzene-1,2-diol
Molecular Weight (g/mol) 186.22
Formula Weight 186.1084
Percent Purity >95