Learn More
rac Desethyl Oxybutynin-d11 Hydrochloride, TRC
Product Code. 30351730
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351730 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30351730 Supplier Toronto Research Chemicals Supplier No. TRCD28947710MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Chemical Name or Material Desethyl Oxybutynin-d11 Hydrochloride
CAS 1216405-15-0
Molecular Formula C20 D11 H16 N O3 . H Cl
Quantity 10 mg
Synonym α-(Cyclohexyl-d11)-α-hydroxybenzeneacetic Acid 4-(Ethylamino)-2-butynyl Ester Hydrochloride Salt
InChI Formula InChI=1S/C20H27NO3.ClH/c1-2-21-15-9-10-16-24-19(22)20(23,17-11-5-3-6-12-17)18-13-7-4-8-14-18;/h3,5-6,11-12,18,21,23H,2,4,7-8,13-16H2,1H3;1H/i4D2,7D2,8D2,13D2,14D2,18D;
SMILES Cl.[2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])(C(O)(C(=O)OCC#CCNCC)c2ccccc2)C([2H])([2H])C1([2H])[2H]
IUPAC Name 4-(ethylamino)but-2-ynyl 2-hydroxy-2-phenyl-2-(1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexyl)acetate;hydrochloride
Molecular Weight (g/mol) 376.96
Formula Weight 376.2448
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.