Learn More
rac-Cotinine-13C,d3, TRC
Specifications
Specifications
| Chemical Name or Material | rac-Cotinine-13C,d3 |
| CAS | 1215842-75-3 |
| Molecular Formula | 13C C9 D3 H9 N2 O |
| Quantity | 25 mg |
| Synonym | (+/-)-1-(Methyl-13C,d3)-5-(2-pyridinyl)-2-pyrrolidinone, (+/-)-Cotinine-13C,d3 |
| InChI Formula | InChI=1S/C10H12N2O/c1-12-9(4-5-10(12)13)8-3-2-6-11-7-8/h2-3,6-7,9H,4-5H2,1H3/i1+1D3 |
| SMILES | [2H][13C]([2H])([2H])N1C(CCC1=O)c2cccnc2 |
| IUPAC Name | 5-(3-pyridyl)-1-(trideuterio(1^{13}C)methyl)pyrrolidin-2-one |
| Molecular Weight (g/mol) | 180.226 |
| Formula Weight | 180.1171 |