missing translation for 'onlineSavingsMsg'
Learn More

rac Bopindolol-d9, TRC

Product Code. 30345146
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30345146 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30345146 Supplier Toronto Research Chemicals Supplier No. TRCB67540210MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac Bopindolol-d9
CAS 1794891-82-9
Molecular Formula C23H19D9N2O3
Quantity 10 mg
Synonym 1-[(1,1-Dimethylethyl-d9)amino]-3-[(2-methyl-1H-indol-4-yl)oxy]-2-propanol 2-Benzoate, (±)-1-[(1,1-Dimethylethyl-d9)amino]-3-[(2-methyl-1H-indol-4-yl)oxy]-2-propanol Benzoate (Ester), (±)-Bopindolol-d9
InChI Formula InChI=1S/C23H28N2O3/c1-16-13-19-20(25-16)11-8-12-21(19)27-15-18(14-24-23(2,3)4)28-22(26)17-9-6-5-7-10-17/h5-13,18,24-25H,14-15H2,1-4H3/i2D3,3D3,4D3
SMILES [2H]C([2H])([2H])C(NCC(COc1cccc2[nH]c(C)cc12)OC(=O)c3ccccc3)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC Name [1-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]amino]-3-[(2-methyl-1H-indol-4-yl)oxy]propan-2-yl] benzoate
Molecular Weight (g/mol) 389.54
Formula Weight 389.2665
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.