Learn More
rac-Arimoclomol Maleic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | rac-Arimoclomol Maleic Acid |
| CAS | 289893-24-9 |
| Molecular Formula | C18H24ClN3O7 |
| Quantity | 10 mg |
| Synonym | N-[2-Hydroxy-3-(1-piperidinyl)propoxy]-3-pyridinecarboximidoyl Chloride 1-Oxide (2Z)-2-Butenedioate |
| InChI Formula | InChI=1S/C14H20ClN3O3.C4H4O4/c15-14(12-5-4-8-18(20)9-12)16-21-11-13(19)10-17-6-2-1-3-7-17;5-3(6)1-2-4(7)8/h4-5,8-9,13,19H,1-3,6-7,10-11H2;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| SMILES | OC(CON=C(Cl)c1ccc[n+]([O-])c1)CN2CCCCC2.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;N-(2-hydroxy-3-piperidin-1-ylpropoxy)-1-oxidopyridin-1-ium-3-carboximidoyl chloride |
| Molecular Weight (g/mol) | 429.85 |
| Formula Weight | 429.1303 |