missing translation for 'onlineSavingsMsg'
Learn More

rac 5-Carboxy Desisopropyl Tolterodine Methyl Ester, TRC

Product Code. 30360591
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360591 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30360591 Supplier Toronto Research Chemicals Supplier No. TRCC17793550MG
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac 5-Carboxy Desisopropyl Tolterodine Methyl Ester
CAS 1391053-28-3
Molecular Formula C20H25NO3
Quantity 50 mg
Synonym 4-Hydroxy-3-[3-[(1-methylethyl)amino]-1-phenylpropyl]-benzoic Acid Methyl Ester
InChI Formula InChI=1S/C20H25NO3/c1-14(2)21-12-11-17(15-7-5-4-6-8-15)18-13-16(20(23)24-3)9-10-19(18)22/h4-10,13-14,17,21-22H,11-12H2,1-3H3
SMILES COC(=O)c1ccc(O)c(c1)C(CCNC(C)C)c2ccccc2
IUPAC Name methyl 4-hydroxy-3-[1-phenyl-3-(propan-2-ylamino)propyl]benzoate
Molecular Weight (g/mol) 327.42
Formula Weight 327.1834
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.