missing translation for 'onlineSavingsMsg'
Learn More

rac-2-(2-Chlorophenyl)-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetonitrile, TRC

Product Code. 30361884
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30361884 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30361884 Supplier Toronto Research Chemicals Supplier No. TRCC379515500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-2-(2-Chlorophenyl)-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetonitrile
CAS 444728-11-4
Molecular Formula C15H13ClN2S
Quantity 500 mg
Synonym alpha-(2-Chlorophenyl)-6,7-dihydro-thieno[3,2-c]pyridine-5(4H)-acetonitrile, (±)-(2-Chlorophenyl)-(6,7-dihydro-4H-thieno[3,2-c]pyrid-5-yl)acetonitrile, 2-(2-Chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetonitrile
InChI Formula InChI=1S/C15H13ClN2S/c16-13-4-2-1-3-12(13)14(9-17)18-7-5-15-11(10-18)6-8-19-15/h1-4,6,8,14H,5,7,10H2
SMILES Clc1ccccc1C(C#N)N2CCc3sccc3C2
IUPAC Name 2-(2-chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetonitrile
Molecular Weight (g/mol) 288.8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.