Learn More
rac 1-Oleoyl Glycerol, TRC
Specifications
Specifications
| Chemical Name or Material | 1-monooleoyl-rac-glycerol |
| CAS | 111-03-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C21 H40 O4 |
| Quantity | 2.5 g |
| Synonym | 9-Octadecenoic acid (Z)-','2,3-dihydroxypropyl ester, Olein','1-mono- (8CI), 1-Glyceryl oleate, 1-Mono(cis-9-octacenoyl)glycerol, 1-Monoolein, 1-Monooleoyl-rac-glycerol, 1-Monooleoyl-rac-glycerol, 1-Monooleoylglycerol, 1-Oleoyl-rac-glycerol, 1-Oleoylglycerol, 1-Oleylglycerol, 2,3-Dihydroxypropyl oleate, Danisco MO 90, GO 991, Glycerin 1-monooleate, Glycerol 1-(9Z-octadecenoate), Glycerol 1-monooleate, Glycerol 1-oleate, Glycerol α-cis-9-octadecenate, Glycerol α-monooleate, Glyceryl monooleate, Nofable GO 991, Rylo MG 19, rac-1-Monoolein, rac-1-Monooleoylglycerol, α-Glyceryl monooloeate, α-Monoolein |
| InChI Formula | InChI=1S/C21H40O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-19-20(23)18-22/h9-10,20,22-23H,2-8,11-19H2,1H3/b10-9- |
| SMILES | CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)CO |
| IUPAC Name | 2,3-dihydroxypropyl (Z)-octadec-9-enoate |
| Molecular Weight (g/mol) | 356.54 |
| Show More |