missing translation for 'onlineSavingsMsg'
Learn More

rac 1-Nitroso-2-methylindoline, TRC

Product Code. 30008039
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008039 100 mg 100mg
30134515 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008039 Supplier Toronto Research Chemicals Supplier No. TRCN529945100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85440-79-5
Molecular Formula C9 H10 N2 O
Molecular Weight (g/mol) 162.19
Synonym (2RS)-2-Methyl-1-nitroso-2,3-dihydro-1H-indole, Indapamide Imp. A (EP), 1H-Indole, 2,3-dihydro-2-methyl-1-nitroso- (9CI, ACI), 1H-Indole, 2,3-dihydro-2-methyl-1-nitroso-, (±)- (ZCI), 2,3-Dihydro-2-methyl-1-nitroso-1H-indole (ACI), 1-Nitroso-2-methylindoline, 2-Methyl-1-nitroso-2,3-dihydro-1H-indole, 2-Methyl-1-nitrosoindoline
IUPAC Name 2-methyl-1-nitroso-2,3-dihydroindole
SMILES CC1Cc2ccccc2N1N=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H10N2O/c1-7-6-8-4-2-3-5-9(8)11(7)10-12/h2-5,7H,6H2,1H3
Formula Weight 162.0793
Percent Purity >95%
Chemical Name or Material (2RS)-2-Methyl-1-nitroso-2,3-dihydro-1H-indole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.