missing translation for 'onlineSavingsMsg'
Learn More

rac 1,2-Diaminopropane-d3, TRC

Product Code. 30367133
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367133 5 mg 5mg
30356948 25 mg 25mg
30350402 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30367133 Supplier Toronto Research Chemicals Supplier No. TRCD4166585MG
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac 1,1,2-Diaminopropane-d3
Molecular Formula C3 D3 H7 N2
Quantity 5 mg
Synonym 1,2-Propanediamine-d3, (-)1,2-Diaminopropane-d3, (RS)-1,2-Propanediamine-d3, 1,2-Propylenediamine-d3, 1-Methyl-1,2-diaminoethane-d3, DL-1,2-Propanediamine-d3, N,N'-Dimethyl-1,2-diaminopropane-d3, NSC 175731-d3
InChI Formula InChI=1S/C3H10N2/c1-3(5)2-4/h3H,2,4-5H2,1H3/i2D2,3D
SMILES [2H]C([2H])(N)C([2H])(C)N
IUPAC Name 1,1,2-trideuteriopropane-1,2-diamine
Molecular Weight (g/mol) 77.14
Formula Weight 77.1032
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.