Learn More
Rabbit IgG control Rabbit, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
53.00€ - 129.00€
Specifications
Antigen | Rabbit IgG control |
---|---|
Applications | Flow Cytometry |
Classification | Polyclonal |
Conjugate | CoraLite 594 |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17791399
|
Proteintech
CL594-30000-20UL |
20 μL |
129.00€
20µL |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17639956
|
Proteintech
CL594-30000-100UL |
100 μL |
53.00€
100µL |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
Description
Rabbit IgG control Polyclonal antibody apecifically detetects Rabbit IgG control antigen in Samples. It is validated for Flow Cytometry.Specifications
Rabbit IgG control | |
Polyclonal | |
Liquid | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
Primary |
Flow Cytometry | |
CoraLite 594 | |
Rabbit | |
IgG | |
Store at -20°C. Avoid exposure to light. |