Learn More
Epredia™ RA Lamb Slide Filing Wallet Cabinet, holds 3 slides
Product Code. 12668556 Shop All Epredia Products
Change view
Click to view available options
Width (Metric) Slide:
126 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Slide unitSize
12668556 126 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12668556 Supplier Epredia™ Supplier No. E48/5
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Store up to three drawers of slide wallets in this slide filing wallet cabinet.
  • Keep your most important slides on hand for pathology, histology and tissue processing lab work
  • Measures 85 x 126mm
Please Note: Availability may vary by country.
TRUSTED_SUSTAINABILITY
Specifications
Type Wallet Cabinet
Device Holds 3 Slides
Height (Metric) Unit 85 mm
Width (Metric) Slide 126 mm