Learn More
Epredia™ RA Lamb Seven-Drawer Filing Cabinets
Shop All Epredia Products
Click to view available options
Color:
Blue
Gray
Green
Slide Capacity:
3000
Unit Size:
Each
Description
- Can be used for the storage of larger microscope slides up to 76 x 51mm
- Can also be used for the storage of paraffin wax blocks in cardboard boxes and Super Mega Cassettes
- It may also be used for the storage of photographic transparencies and most grid boxes used in electron microscopy
- If a drawer is fitted with the Raymond A Lamb divider, two rows of 76 x 26 slides can be stored
- Capacity of 1500 or 3000 slides depending on model
Specifications
Specifications
| Type | Slide Filing Cabinet |
| Slide Capacity | 3000 |
| Color | Green |
| Device Holds | 3000 Slides |
| Length (Metric) Slide | 76 mm |
| Material | Cardboard Box, Steel, Paraffin Wax Block |
| Width (Metric) Slide | 26/51 mm |