Learn More
Thermo Scientific™ RA Lamb Embryo Dishes, 30mm
Product Code. 12678586 Shop All Epredia Products
Change view
Click to view available options
Diameter (Metric):
30 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
12678586 30 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12678586 Supplier Thermo Scientific™ Supplier No. E90
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
View or stain free-floating specimens with Thermo Scientific™ RA Lamb Embryo Dishes, molded of clear glass with glass lids for viewing.
TRUSTED_SUSTAINABILITY
Specifications
Description 30mm
Material Glass
Type Embryo Dish
Diameter (Metric) 30 mm