Learn More
Epredia™ RA Lamb BlockStore Storage Options for the LabStack Storage System
Product Code. p-4520721 Shop All Epredia Products
Click to view available options
Product Type:
8-Drawer Cabinet
Color:
Gray
Unit Size:
Each
Product Code. 12181830
Brand: Epredia™ E43/22GREY
Store lab supplies that don't typically fit in standard-size compartments with this versatile, expandable system.
  • Compatible with all standard LabStack cabinets and options
  • 8-Drawer Cabinet comes with two rows of four drawers
  • Each drawer has enough dividers to produce seven rows 1.2 in. (30mm) wide
  • Accepts embedding rings, processing cassettes, and unmounted paraffin blocks
TRUSTED_SUSTAINABILITY
Specifications
Product Type 8-Drawer Cabinet
Color Gray