missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ RA 2
GREENER_CHOICE

Product Code. 18797326
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18797326

Brand: Tocris Bioscience™ 7129/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent negative modulator of KCa2.3 and KCa3.1 channels

RA 2 is a potent negative modulator of calcium-activated potassium channels KCa2.3 and KCa3.1 (IC50 values are 2 and 17 nM, respectively), which displays >55-fold selectively for KCa2 and KCa3.1 over other KCa, Kv and Kir channels. In ex vivo porcine coronary endothelium, RA 2 inhibits SKA 31 (Cat. No. 3670)-induced KCa2/KCa3.1 currents and TRAM 34 (Cat. No. 2946)-insensitive KCa2 currents. Reduces heart rate in mice in a KCa3.1-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Phenylenebis(methylene)bis(3-fluoro-4-hydroxybenzoate)
CAS 1867107-62-7
Quantity 10 mg
Target Ca2+-Activated Potassium Channel Modulators
Molecular Formula C22H16F2O6
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.